ENA387772326 Enamine

(2R)-2-aminopropanenitrile hydrochloride

  • MDL number MFCD22628484
  • Cas Number : 1385022-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120035
Availability details for
Quantity
  • Availability
  • Add to Cart