ENA387772328 Enamine

ethyl 4-amino-3-methylbutanoate hydrochloride

  • MDL number MFCD22628486
  • Cas Number : 204503-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120059

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120059
Availability details for
Quantity
  • Availability
  • Add to Cart