ENA387772331 Enamine

6-(pyridin-3-yloxy)pyridin-3-amine hydrochloride

  • MDL number MFCD22628489
  • Cas Number : 1423024-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120072

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120072
Availability details for
Quantity
  • Availability
  • Add to Cart