ENA387772342 Enamine

2-amino-3-methoxypropanamide hydrochloride

  • MDL number MFCD22628503
  • Cas Number : 17463-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120170
Availability details for
Quantity
  • Availability
  • Add to Cart