ENA387772344 Enamine

[5-(chlorodifluoromethyl)-1,2,4-oxadiazol-3-yl]methanamine hydrochloride

  • MDL number MFCD22628506
  • Cas Number : 1423033-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-120235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120235
Availability details for
Quantity
  • Availability
  • Add to Cart