ENA387772347 Enamine

methyl 2-amino-2-(3-hydroxyphenyl)acetate hydrochloride

  • MDL number MFCD22628509
  • Cas Number : 179814-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120257

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120257
Availability details for
Quantity
  • Availability
  • Add to Cart