ENA387772352 Enamine

(2S)-2-amino-N-benzyl-N-ethyl-4-methylpentanamide hydrochloride

  • MDL number MFCD22628515
  • Cas Number : 1423015-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-120300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120300
Availability details for
Quantity
  • Availability
  • Add to Cart