ENA387772353 Enamine

(2S)-2-amino-N-benzyl-N,3-dimethylpentanamide hydrochloride

  • MDL number MFCD22628516
  • Cas Number : 1423017-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120301

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120301
Availability details for
Quantity
  • Availability
  • Add to Cart