ENA387772354 Enamine

1-[(1-benzyl-1H-1,2,3,4-tetrazol-5-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD22628517
  • Cas Number : 1423026-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120302

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120302
Availability details for
Quantity
  • Availability
  • Add to Cart