ENA387772361 Enamine

2-(5-chloro-1-methyl-1H-imidazol-2-yl)-2,2-difluoroacetic acid hydrochloride

  • MDL number MFCD22628526
  • Cas Number : 1423023-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120406
Availability details for
Quantity
  • Availability
  • Add to Cart