ENA387772368 Enamine

5-(aminomethyl)-1-benzylpyrrolidin-3-ol

  • MDL number MFCD22628538
  • Cas Number : 1423023-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120530

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120530
Availability details for
Quantity
  • Availability
  • Add to Cart