ENA387772375 Enamine

3-(morpholin-2-yl)benzonitrile hydrochloride

  • MDL number MFCD22628545
  • Cas Number : 1423033-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120596

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-120596
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart