ENA387772383 Enamine

4-methoxy-2-(propane-1-sulfonamido)benzene-1-sulfonyl chloride

  • MDL number MFCD22628556
  • Cas Number : 1423033-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-120739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-120739
Availability details for
Quantity
  • Availability
  • Add to Cart