ENA387772394 Enamine

2-(pyridin-2-yl)-2,3-dihydro-1H-isoindol-4-amine

  • MDL number MFCD10007264
  • Cas Number : 1017388-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119144
Availability details for
Quantity
  • Availability
  • Add to Cart