ENA387772396 Enamine

1-[(3-bromophenyl)methyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00836294
  • Cas Number : 313376-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119146
Availability details for
Quantity
  • Availability
  • Add to Cart