ENA387772400 Enamine

3-phenylcycloheptan-1-one

  • MDL number MFCD16659524
  • Cas Number : 74395-08-7
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 17-18
Iupac Name 3-phenylcycloheptanone
Inchi Code 1S/C13H16O/c14-13-9-5-4-8-12(10-13)11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10H2
Inchi Key JLDCLDZGNYDRKX-UHFFFAOYSA-N

Description

Melting Point 17-18
Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-119151

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-119151
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart