ENA387772403 Enamine

1-(3-bromopyridin-2-yl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD14651587
  • Cas Number : 1249324-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119154

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119154
Availability details for
Quantity
  • Availability
  • Add to Cart