ENA387772411 Enamine

1-phenyl-3-(1,3-thiazol-4-yl)propan-2-amine

  • MDL number MFCD16858352
  • Cas Number : 1341072-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119163

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119163
Availability details for
Quantity
  • Availability
  • Add to Cart