ENA387772412 Enamine

2-chloro-5,6,7,8-tetrahydroquinolin-8-one

  • MDL number MFCD13189689
  • Cas Number : 129337-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-129
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119164
Availability details for
Quantity
  • Availability
  • Add to Cart