ENA387772425 Enamine

2-[N-(butan-2-yl)2-bromobenzenesulfonamido]acetic acid

  • MDL number MFCD12525663
  • Cas Number : 1183039-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119184
Availability details for
Quantity
  • Availability
  • Add to Cart