ENA387772428 Enamine

methyl 1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD22628348
  • Cas Number : 89032-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-54
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112510

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-112510
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart