ENA387772432 Enamine

methyl 5-(iodomethyl)-2-methyl-4,5-dihydrofuran-3-carboxylate

  • MDL number MFCD22628349
  • Cas Number : 128738-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113073
Availability details for
Quantity
  • Availability
  • Add to Cart