ENA387772434 Enamine

2-amino-4-methylheptanoic acid

  • MDL number MFCD09914815
  • Cas Number : 101257-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119200
Availability details for
Quantity
  • Availability
  • Add to Cart