ENA387772436 Enamine

3-(4-fluorophenyl)pyrrolidin-3-ol hydrochloride

  • MDL number MFCD01687444
  • Cas Number : 2248-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119224
Availability details for
Quantity
  • Availability
  • Add to Cart