ENA387772437 Enamine

5-methylnaphthalene-1-carboxylic acid

  • MDL number MFCD18415107
  • Cas Number : 4527-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-188
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119225
Availability details for
Quantity
  • Availability
  • Add to Cart