ENA387772441 Enamine

2-methyl-5-(1,3-oxazol-2-yl)aniline

  • MDL number MFCD14657173
  • Cas Number : 1249267-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-119229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119229
Availability details for
Quantity
  • Availability
  • Add to Cart