ENA387772448 Enamine

N-[(3-aminophenyl)methyl]-N-ethylpyridine-3-carboxamide

  • MDL number MFCD12579089
  • Cas Number : 1154321-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119242
Availability details for
Quantity
  • Availability
  • Add to Cart