ENA387772450 Enamine

2-methyl-2-(methylamino)propanoic acid hydrochloride

  • MDL number MFCD06795786
  • Cas Number : 1223405-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119244
s_phrases_eu 26
Availability details for
Quantity
  • Availability
  • Add to Cart