ENA387772470 Enamine

3-{1-[(tert-butoxy)carbonyl]pyrrolidin-3-yl}propanoic acid

  • MDL number MFCD08234795
  • Cas Number : 885271-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114316
Availability details for
Quantity
  • Availability
  • Add to Cart