ENA387772473 Enamine

5-fluoro-4-methyl-1,3-thiazol-2-amine

  • MDL number MFCD18816988
  • Cas Number : 947179-49-9
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-4-methyl-1,3-thiazol-2-amine
Inchi Code 1S/C4H5FN2S/c1-2-3(5)8-4(6)7-2/h1H3,(H2,6,7)
Inchi Key QBDZOLWTYRZKBV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-119316

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-119316
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart