ENA387772478 Enamine

4-bromo-6-chloro-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD18208167
  • Cas Number : 1260017-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119328
Availability details for
Quantity
  • Availability
  • Add to Cart