ENA387772479 Enamine

3-[(2-methoxyphenyl)methoxy]-2-methylaniline

  • MDL number MFCD16497385
  • Cas Number : 1282185-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119330
Availability details for
Quantity
  • Availability
  • Add to Cart