ENA387772480 Enamine

6-bromo-4-chloro-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD18207830
  • Cas Number : 1260017-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124...126
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119331

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119331
Availability details for
Quantity
  • Availability
  • Add to Cart