ENA387772485 Enamine

2-bromo-4-chloropyridin-3-amine

  • MDL number MFCD13193358
  • Cas Number : 357263-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119339
Availability details for
Quantity
  • Availability
  • Add to Cart