ENA387772491 Enamine

2-[2-(4-chlorophenyl)-1,3-thiazol-4-yl]ethan-1-amine hydrochloride

  • MDL number MFCD12963845
  • Cas Number : 1187830-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119347

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119347
Availability details for
Quantity
  • Availability
  • Add to Cart