ENA387772493 Enamine

1-(6-methylpyrimidin-4-yl)-1H-pyrazol-5-amine

  • MDL number MFCD16156313
  • Cas Number : 1247244-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119349

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119349
Availability details for
Quantity
  • Availability
  • Add to Cart