ENA387772496 Enamine

(3-methanesulfonylpropyl)(methyl)amine

  • MDL number MFCD12778857
  • Cas Number : 373356-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119352

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-119352
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart