ENA387772500 Enamine

(1S)-1-(4-ethylphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD16295041
  • Cas Number : 212968-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119358

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119358
Availability details for
Quantity
  • Availability
  • Add to Cart