ENA387772504 Enamine

2-(oxan-4-yloxy)-2-phenylacetic acid

  • MDL number MFCD13332085
  • Cas Number : 1306175-72-3
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115071
Availability details for
Quantity
  • Availability
  • Add to Cart