ENA387772507 Enamine

tert-butyl 3-hydroxy-3-[(pyridin-4-yl)methyl]azetidine-1-carboxylate

  • MDL number MFCD20864850
  • Cas Number : 1423032-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119367
Availability details for
Quantity
  • Availability
  • Add to Cart