ENA387772510 Enamine

2-(1,5-dimethyl-1H-pyrrol-2-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD22628350
  • Cas Number : 1423026-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115238

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115238
Availability details for
Quantity
  • Availability
  • Add to Cart