ENA387772511 Enamine

N,N-diethyl-4-methylpiperazine-1-carboxamide

  • MDL number MFCD00023288
  • Cas Number : 90-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119374
Availability details for
Quantity
  • Availability
  • Add to Cart