ENA387772515 Enamine

(dimethyl-1,3-thiazol-4-yl)methanamine

  • MDL number MFCD22628410
  • Cas Number : 1423032-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119412
Availability details for
Quantity
  • Availability
  • Add to Cart