ENA387772524 Enamine

2-chloro-5-(methylamino)pyridine-4-carboxylic acid

  • MDL number MFCD15528057
  • Cas Number : 945737-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119428
Availability details for
Quantity
  • Availability
  • Add to Cart