ENA387772525 Enamine

3-cyclohexyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD17193820
  • Cas Number : 1037061-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119430
Availability details for
Quantity
  • Availability
  • Add to Cart