ENA387772534 Enamine

4-chloro-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

  • MDL number MFCD09880128
  • Cas Number : 1000340-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119444

Safety Information

msds https://www.enaminestore.com/catalog/EN300-119444
Availability details for
Quantity
  • Availability
  • Add to Cart