ENA387772535 Enamine

ethyl 4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD02647051

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-151
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-119447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119447
Availability details for
Quantity
  • Availability
  • Add to Cart