ENA387772536 Enamine

2-ethyl-6-phenoxy-1,4-dihydroquinolin-4-one

  • MDL number MFCD13762639
  • Cas Number : 1155599-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115745
Availability details for
Quantity
  • Availability
  • Add to Cart