ENA387772538 Enamine

potassium trifluoro({4-[(oxan-4-yl)methoxy]phenyl})boranuide

  • MDL number MFCD18605946

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119457
Availability details for
Quantity
  • Availability
  • Add to Cart