ENA387772553 Enamine

4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-1,2-dihydropyrimidin-2-one

  • MDL number MFCD00870151
  • Cas Number : 143491-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-119501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119501
Availability details for
Quantity
  • Availability
  • Add to Cart